missing translation for 'onlineSavingsMsg'
Learn More

4-(Pentafluorothio)phenol, 97%

Product Code. p-7032080
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15416227

Brand: Thermo Scientific Alfa Aesar H33413.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 774-94-7 | C6H5F5OS | 220.16 g/mol

Synthesis of hexylamides using fluorine derivatives as intermediate active esters. Various non-metallic derivatives of pentafluorothiophenol, containing sulfur bonded to boron, carbon, silicon, germanium, phosphorus, arsenic, antimony, sulfur, and chlorine, can be prepared.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of hexylamides using fluorine derivatives as intermediate active esters. Various non-metallic derivatives of pentafluorothiophenol, containing sulfur bonded to boron, carbon, silicon, germanium, phosphorus, arsenic, antimony, sulfur, and chlorine, can be prepared.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 774-94-7
Molecular Formula C6H5F5OS
Molecular Weight (g/mol) 220.16
MDL Number MFCD03788516
InChI Key XHJLGVIUMCBMHL-UHFFFAOYSA-N
Synonym 4-pentafluorothio phenol, 4-pentafluorosulfanyl phenol, 4-hydroxyphenylsulfur pentafluoride, 4-pentafluoro-$l^ 6-sulfanyl phenol, 4-pentafluoro-lambda∼6∼-sulfanyl phenol, sulfur,pentafluoro 4-hydroxyphenyl-, oc-6-21-9ci, 4-pentafluorosulfanylphenol, 4-pentafluorosulfur phenol, 4-pentafluorosulphanyl phenol, 4-pentafluorothio vi phenol
PubChem CID 2779203
IUPAC Name 4-(pentafluoro-$l^{6}-sulfanyl)phenol
SMILES OC1=CC=C(C=C1)S(F)(F)(F)(F)F

Specifications

Quantity 1 g
Formula Weight 220.16
Percent Purity 97%
Chemical Name or Material 4-(Pentafluorothio)phenol
compliance-icons
Product Identifier
  • 4-(Pentafluorothio)phenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 4-Hydroxyphenylsulfur pentafluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.