missing translation for 'onlineSavingsMsg'
Learn More

4-(p-Iodophenyl)butyric Acid, TRC

Product Code. 30010551
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010551 1 g 1g
30005165 10 g 10g
30005711 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010551 Supplier Toronto Research Chemicals Supplier No. TRCI7175401G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27913-58-2
Molecular Formula C10 H11 I O2
Molecular Weight (g/mol) 290.1
Synonym 4-Iodo-benzenebutanoic Acid, Butyric acid, γ-(p-iodophenyl)- (4CI), 4-(4-Iodophenyl)butanoic acid, 4-(4-Iodophenyl)butyric acid, 4-(p-Iodophenyl)butanoic acid
IUPAC Name 4-(4-iodophenyl)butanoic acid
SMILES OC(=O)CCCc1ccc(I)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H11IO2/c11-9-6-4-8(5-7-9)2-1-3-10(12)13/h4-7H,1-3H2,(H,12,13)
Formula Weight 289.9804
Chemical Name or Material 4-(p-Iodophenyl)butyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.