missing translation for 'onlineSavingsMsg'
Learn More

4-(p-Chloro-alpha-phenylbenzyl)-1-piperazineethanol, TRC

Product Code. 30006810
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006810 1 g 1g
30009508 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006810 Supplier Toronto Research Chemicals Supplier No. TRCC3775651G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109806-71-5
Molecular Formula C19 H23 Cl N2 O
Molecular Weight (g/mol) 330.85
Synonym 1-Piperazineethanol, 4-[(4-chlorophenyl)phenylmethyl]-, 1-Piperazineethanol, 4-(p-chloro-α-phenylbenzyl)- (6CI), 4-[(4-Chlorophenyl)phenylmethyl]-1-piperazineethanol, 2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethanol, 2-[4-[(RS)-(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethanol
IUPAC Name 2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethanol
SMILES OCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H23ClN2O/c20-18-8-6-17(7-9-18)19(16-4-2-1-3-5-16)22-12-10-21(11-13-22)14-15-23/h1-9,19,23H,10-15H2
Formula Weight 330.1499
Percent Purity >95%
Chemical Name or Material 4-(p-Chloro-alpha-phenylbenzyl)-1-piperazineethanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.