missing translation for 'onlineSavingsMsg'
Learn More

4-(p-Carboxyphenyl)-1,2,4-triazole, 97%, Thermo Scientific™

Product Code. 15542303 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15542303 1 g 1g
15552303 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15542303 Supplier Thermo Scientific Acros Supplier No. 449960010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 157069-48-2
Molecular Formula C9H7N3O2
Molecular Weight (g/mol) 189.17
InChI Key QCNDLFQKKMPZKH-UHFFFAOYSA-N
Synonym 4-4h-1,2,4-triazol-4-yl benzoic acid, 4-1,2,4-triazol-4-yl benzoic acid, 4-1,2,4 triazol-4-yl-benzoic acid, benzoic acid, 4-4h-1,2,4-triazol-4-yl, 4-4-carboxyphenyl-1,2,4-triazole, 4-4-carboxyphenyl-4h-1,2,4-triazole, 4-4h-1,2,4-triazol-4-yl-benzoic acid
PubChem CID 3163581
IUPAC Name 4-(1,2,4-triazol-4-yl)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)N2C=NN=C2

Specifications

Melting Point 330.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 4-(p-Carboxyphenyl)-1,2,4-triazole
compliance-icons
Product Identifier
  • 4-(p-Carboxyphenyl)-1,2,4-triazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.