missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-4-(2-thienyl)butyric acid, 98%, Thermo Scientific™

Product Code. 11332026
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332026 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11332026 Supplier Thermo Scientific Alfa Aesar Supplier No. L03100.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4653-08-1
Molecular Formula C8H8O3S
Molecular Weight (g/mol) 184.21
MDL Number MFCD00022502
InChI Key ULJMYWHLMLRYSO-UHFFFAOYSA-N
Synonym 3-2-thenoyl propionic acid, 4-oxo-4-thiophen-2-yl butanoic acid, 4-oxo-4-2-thienyl butanoic acid, 4-oxo-4-2-thienyl butyric acid, 3-2-thenoyl propanoic acid, 4-thien-2-yl-4-oxobutyric acid, 4-oxo-4-thien-2-yl butyric acid, 4-thien-2-yl-4-oxobutanoic acid, 4-oxo-4-thien-2-yl butanoic acid, 4-oxo-4-thiophen-2-yl-butyric acid
PubChem CID 78385
ChEBI CHEBI:64434
IUPAC Name 4-oxo-4-thiophen-2-ylbutanoic acid
SMILES C1=CSC(=C1)C(=O)CCC(=O)O

Specifications

Melting Point 116°C to 121°C
Odor Odorless
Quantity 5 g
Beilstein 133870
Formula Weight 184.21
Percent Purity 98%
Chemical Name or Material 4-Oxo-4-(2-thienyl)butyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.