missing translation for 'onlineSavingsMsg'
Learn More

4-oxo-4-(2-thienyl)butanoic acid, Thermo Scientific™

Product Code. 10774277
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 4653-08-1
Molecular Formula C8H8O3S
Molecular Weight (g/mol) 184.21
InChI Key ULJMYWHLMLRYSO-UHFFFAOYSA-N
Synonym 3-2-thenoyl propionic acid, 4-oxo-4-thiophen-2-yl butanoic acid, 4-oxo-4-2-thienyl butanoic acid, 4-oxo-4-2-thienyl butyric acid, 3-2-thenoyl propanoic acid, 4-thien-2-yl-4-oxobutyric acid, 4-oxo-4-thien-2-yl butyric acid, 4-thien-2-yl-4-oxobutanoic acid, 4-oxo-4-thien-2-yl butanoic acid, 4-oxo-4-thiophen-2-yl-butyric acid
PubChem CID 78385
ChEBI CHEBI:64434
IUPAC Name 4-oxo-4-thiophen-2-ylbutanoic acid
SMILES C1=CSC(=C1)C(=O)CCC(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 25 g
Formula Weight 184.21
Percent Purity 97%
Chemical Name or Material 4-Oxo-4-(2-thienyl)butanoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'