missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-4-(2-thienyl)butanoic acid 97%, ACROS Organics™

Product Code. 10657674 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10657674 25 g Glass bottle 25g
10177843 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10657674 Supplier Thermo Scientific Acros Supplier No. 124500250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4653-08-1
Molecular Formula C8H8O3S
Molecular Weight (g/mol) 184.21
MDL Number MFCD00022502
InChI Key ULJMYWHLMLRYSO-UHFFFAOYSA-N
Synonym 3-2-thenoyl propionic acid, 4-oxo-4-thiophen-2-yl butanoic acid, 4-oxo-4-2-thienyl butanoic acid, 4-oxo-4-2-thienyl butyric acid, 3-2-thenoyl propanoic acid, 4-thien-2-yl-4-oxobutyric acid, 4-oxo-4-thien-2-yl butyric acid, 4-thien-2-yl-4-oxobutanoic acid, 4-oxo-4-thien-2-yl butanoic acid, 4-oxo-4-thiophen-2-yl-butyric acid
PubChem CID 78385
ChEBI CHEBI:64434
IUPAC Name 4-oxo-4-thiophen-2-ylbutanoic acid
SMILES C1=CSC(=C1)C(=O)CCC(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 118°C to 121°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Formula Weight 184.21
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 4-Oxo-4-(2-thienyl)butanoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.