missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-4-(1-pyrenyl)butyric acid, 97%, Thermo Scientific™

Product Code. 15254764 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15254764 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15254764 Supplier Thermo Scientific Acros Supplier No. 449330050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7499-60-7
Melting Point 182.0°C to 184.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C20H14O3
MDL Number MFCD00010369
Quantity 5 g
Synonym 4-oxo-4-pyren-1-yl-butyric acid, 4-oxo-4-pyren-1-yl butanoic acid, 1-pyrenebutanoic acid, .gamma.-oxo, gamma-oxopyrene-1-butyric acid, 4-oxo-4-pyren-1-ylbutyric acid, 1-pyrenebutyric acid, .gamma.-oxo, 4-oxo-4-pyrenylbutanoic acid, 1-pyrenebutanoic acid,, a-oxo, 4-oxo-4-pyren-1-yl-butyricacid, pubchem23448
Solubility Information Solubility in water: insoluble.
InChI Key AAJQTWWVKREOQP-UHFFFAOYSA-N
SMILES OC(=O)CCC(=O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
IUPAC Name 4-oxo-4-pyren-1-ylbutanoic acid
Molecular Weight (g/mol) 302.33
PubChem CID 82011
Formula Weight 302.32
Percent Purity 97%
Chemical Name or Material 4-Oxo-4-(1-pyrenyl)butyric acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Oxo-4(1-pyrenyl)butyric acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.