missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-2,2,6,6-tetramethylpiperidinooxy, 95%, free radical, Thermo Scientific™

Product Code. p-177879 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10275990

Brand: Thermo Scientific Acros 176860010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Polymerization reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2896-70-0
Molecular Formula C9H16NO2
Molecular Weight (g/mol) 170.23
MDL Number MFCD00006187
Synonym 2,2,6,6-Tetramethyl-4-oxopiperidinooxy, 4-Oxo-TEMPO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 33°C to 38°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 170.23
Percent Purity 95%
Physical Form Crystalline Powder, Crystals or Needles
Chemical Name or Material 4-Oxo-2,2,6,6-tetramethylpiperidinooxy, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.