missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Oxo-2,2,6,6-tetramethylpiperidine-d17, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.50mg
Specifications
Specifications
| Chemical Name or Material | 4-Oxo-2,2,6,6-tetramethylpiperidine-d17 |
| CAS | 52168-48-6 |
| Molecular Formula | C9 D17 N O |
| Quantity | 2.5 mg |
| Synonym | 4-Piperidinone-1,3,3,5,5-d5','2,2,6,6-tetra(methyl-d3)- (9CI','ACI), 2,2,6,6-Tetra(methyl-d3)-4-piperidinone-1,3,3,5,5-d5 (ACI), 2,2,6,6-Tetramethyl-4-oxo-1-piperidinyloxy-d16, Perdeuterated 2,2,6,6-tetramethyl-4-piperidone, 4-Oxo-2,2,6,6-tetramethylpiperidine-d17, 2,2,6,6-Tetramethyl-4-piperidone-D17 |
| InChI Formula | InChI=1S/C9H17NO/c1-8(2)5-7(11)6-9(3,4)10-8/h10H,5-6H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2/hD |
| SMILES | [2H]N1C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])C(=O)C([2H])([2H])C1(C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC Name | 1,3,3,5,5-pentadeuterio-2,2,6,6-tetrakis(trideuteriomethyl)piperidin-4-one |
| Molecular Weight (g/mol) | 172.3421 |
| Formula Weight | 172.2377 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction