missing translation for 'onlineSavingsMsg'
Learn More

4-Octyne, 98+%, ACROS Organics™

Brand:  Acros Organics 223520100

Code : 29

Additional Details : CAS Number : 1942-45-6 Weight : 0.06000kg Transport : UN number : 1993 Chem class : 3 Pack group : III


Packaging Glass bottle
Quantity 10mL

 View more versions of this product

Product Code. 10072920

  • £30.33 / 10mL

Estimated Shipment
05-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1942-45-6
110.2
GZTNBKQTTZSQNS-UHFFFAOYSA-N
16029
CCCC#CCCC
C8H14
MFCD00009471
4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
oct-4-yne

Specifications

4-Octyne
29°C
Glass bottle
0.75
Colorless to Yellow
10mL
98.5
98% min. (GC)
CH3CH2CH2C≡CCH2CH2CH3
01, III, 1006
GZTNBKQTTZSQNS-UHFFFAOYSA-N
oct-4-yne
16029
Liquid
0.7500g/mL
Authentic
1.4240 to 1.4260
131.0°C to 132.0°C
-103.0°C
1942-45-6
100.0
C8H14
MFCD00009471
4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
CCCC#CCCC
110.2
110.2
98+%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Octyne

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS Documents