missing translation for 'onlineSavingsMsg'
Learn More

4-Octyne, 98+%

Product Code. 10117463 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10117463 50 mL Glass bottle 50mL
10072920 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10117463 Supplier Thermo Scientific Acros Supplier No. 223520500
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1942-45-6 | C8H14 | 110.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1942-45-6
Molecular Formula C8H14
Molecular Weight (g/mol) 110.2
MDL Number MFCD00009471
InChI Key GZTNBKQTTZSQNS-UHFFFAOYSA-N
Synonym 4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
PubChem CID 16029
IUPAC Name oct-4-yne
SMILES CCCC#CCCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -103.0°C
Color Colorless to Yellow
Density 0.7500g/mL
Boiling Point 131.0°C to 132.0°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2C≡CCH2CH2CH3
Refractive Index 1.4240 to 1.4260
Quantity 50 mL
Beilstein 01, III, 1006
Specific Gravity 0.75
Formula Weight 110.2
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 4-Octyne
Show More Show Less
compliance-icons
Product Identifier
  • 4-Octyne
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.