missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ifenprodil tartrate, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Ifenprodil tartrate is a typical noncompetitive NMDA receptor antagonist. Ifenprodil tartrate exerts high affinity at NR1A/NR2B receptors (IC50=0.34 μM) over 400-fold than at NR1A/NR2A receptors (IC50=146 μM). Ifenprodil tartrate inhibits GIRK (Kir3), reduces inward currents through the basal GIRK activity. Ifenprodil tartrate has the potential to be a cerebral vasodilator.
Specifications
Specifications
| Chemical Name or Material | Ifenprodil tartrate |
| CAS | 23210-58-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H27NO2.1/2C4H6O6 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 500 mg/mL (1248.47 mM) ∣H2O : 5 mg/mL (12.48 mM; Need ultrasonic) |
| SMILES | OC(C1=CC=C(O)C=C1)C(C)N(CC2)CCC2CC3=CC=CC=C3.O=C(O)[C@H](O)[C@@H](O)C(O)=O.[1/2] |
| Molecular Weight (g/mol) | 400.49 |
| Formula Weight | 400.49 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction