missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 4-Octyl Itaconate
GREENER_CHOICE

Product Code. 18799256
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18799256

Brand: Tocris Bioscience™ 6662/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nrf2 activator; cell permeable

4-Octyl itaconate activates Nrf2. Inhibits LPS-induced increases in IL-1β mRNA, HIF-1α and IL-10 in macrophages. Decreases cytokine production in response to LPS in mice and prolongs survival. Cell permeable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methylenebutanedioic acid 4-octyl ester
CAS 3133-16-2
Quantity 10 mg
Molecular Formula C13H22O4
Formula Weight 242.31
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.