missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 4-Octyl Itaconate

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
4-Octyl itaconate activates Nrf2. Inhibits LPS-induced increases in IL-1β mRNA, HIF-1α and IL-10 in macrophages. Decreases cytokine production in response to LPS in mice and prolongs survival. Cell permeable.
Specifications
Specifications
| Chemical Name or Material | 2-Methylenebutanedioic acid 4-octyl ester |
| CAS | 3133-16-2 |
| Quantity | 50 mg |
| Molecular Formula | C13H22O4 |
| Formula Weight | 242.31 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction