missing translation for 'onlineSavingsMsg'
Learn More

4-O-Methyl honokiol, MedChemExpress

Product Code. 16452658
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16452658

Brand: Medchem Express HYU00450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 4-O-Methyl honokiol is a natural neolignan isolated from Magnolia officinalis, acts as a PPARγ agonist, and inhibtis NF-κB activity, used for cancer and inflammation research.

4-O-Methyl honokiol is a natural neolignan isolated from Magnolia officinalis, acts as a PPARγ agonist, and inhibtis NF-κB activity, used for cancer and inflammation research.

Specifications

Chemical Name or Material 4-O-Methyl honokiol
CAS 68592-15-4
Color Orange Red
Purity Grade Notes Research
Molecular Formula C19H20O2
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (356.68 mM; Need ultrasonic)
SMILES OC1=CC=C(CC=C)C=C1C2=CC=C(OC)C(CC=C)=C2
Molecular Weight (g/mol) 280.36
Formula Weight 280.36
Percent Purity 99.47%
Grade Research
Physical Form Liquid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.