missing translation for 'onlineSavingsMsg'
Learn More

4-O-Methyl-D-glucuronic Acid, TRC

Product Code. 30179255
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179255 2.5 mg 2.5mg
30179059 1 mg 1mg
30179048 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179255 Supplier Toronto Research Chemicals Supplier No. TRCM3228082.5MG
 Restricted Item
Estimated Shipment: 15-10-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 2463-49-2
Molecular Formula C7 H12 O7
Molecular Weight (g/mol) 208.17
Synonym 4-O-Methylglucuronic Acid
IUPAC Name (2S,3S,4R,5R)-2,4,5-trihydroxy-3-methoxy-6-oxohexanoic acid
SMILES CO[C@@H]([C@H](O)[C@@H](O)C=O)[C@H](O)C(=O)O

Specifikationer

Quantity 2.5 mg
InChI Formula InChI=1S/C7H12O7/c1-14-6(5(11)7(12)13)4(10)3(9)2-8/h2-6,9-11H,1H3,(H,12,13)/t3-,4+,5-,6-/m0/s1
Formula Weight 208.0583
Chemical Name or Material 4-O-Methyl-D-glucuronic Acid linear
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.