missing translation for 'onlineSavingsMsg'
Learn More

(±)-4-O-Ethyl Albuterol, TRC

Product Code. 30383664
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383664 25 mg
30371989 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383664 Supplier Toronto Research Chemicals Supplier No. TRCE67838025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2387728-91-6
Molecular Formula C15H25NO3
Molecular Weight (g/mol) 267.36
Synonym 2-(tert-Butylamino)-1-(4-ethoxy-3-(hydroxymethyl)phenyl)ethanol
SMILES OCC1=C(OCC)C=CC(C(O)CNC(C)(C)C)=C1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H25NO3/c1-5-19-14-7-6-11(8-12(14)10-17)13(18)9-16-15(2,3)4/h6-8,13,16-18H,5,9-10H2,1-4H3
Chemical Name or Material (±)-4-O-Ethyl Albuterol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.