missing translation for 'onlineSavingsMsg'
Learn More

4-O-beta-Glucopyranosyl-cis-coumaric alphacid (>80%), TRC

Product Code. 30393990
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393990 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393990 Supplier Toronto Research Chemicals Supplier No. TRCG4190401MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-O-beta-Glucopyranosyl-cis-coumaric alphacid
CAS 117405-48-8
Molecular Formula C15H18O8
Quantity 1 mg
Synonym (2Z)-3-[4-(β-D-Glucopyranosyloxy)phenyl]-2-propenoic Acid, (Z)-p-Coumaric Acid 4-O-β-D-glucopyranoside, 4-O-β-D-Glucopyranosyl-cis-p-coumaric Acid, cis-p-Coumaric Acid 4-O-β-D-glucopyranoside
InChI Formula InChI=1S/C15H18O8/c16-7-10-12(19)13(20)14(21)15(23-10)22-9-4-1-8(2-5-9)3-6-11(17)18/h1-6,10,12-16,19-21H,7H2,(H,17,18)/b6-3-/t10-,12-,13+,14-,15?/m1/s1
SMILES OC(/C=C\C1=CC=C(OC2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)C=C1)=O
IUPAC Name (Z)-3-[4-[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]prop-2-enoic acid
Molecular Weight (g/mol) 326.3
Formula Weight 326.1002
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.