missing translation for 'onlineSavingsMsg'
Learn More

4-O-Benzyl-D-glucal, 97%, Thermo Scientific Chemicals

Product Code. 15407168
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15407168 1 g 1g
15417168 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15407168 Supplier Thermo Scientific Alfa Aesar Supplier No. H60678.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58871-11-7
Molecular Formula C13H16O4
Molecular Weight (g/mol) 236.27
MDL Number MFCD01863610
InChI Key OIWQZGBSQDJVJN-UPJWGTAASA-N
Synonym 4-o-benzyl-d-glucal, 2r,3s,4r-3-benzyloxy-2-hydroxymethyl-3,4-dihydro-2h-pyran-4-ol, 4-o-benzyl-d-glucal,, 1,2-dideoxy-4-o-benzyl-d-arabino-1-hexenopyranose, 2r,3s,4r-2-hydroxymethyl-3-phenylmethoxy-3,4-dihydro-2h, 2r,3s,4r-2-hydroxymethyl-3-phenylmethoxy-3,4-dihydro-2h-pyran-4-ol, 5s,4r,6r-6-hydroxymethyl-5-phenylmethoxy-4h-5,6-dihydropyran-4-ol
PubChem CID 2734727
IUPAC Name (2R,3S,4R)-2-(hydroxymethyl)-3-phenylmethoxy-3,4-dihydro-2H-pyran-4-ol
SMILES OC[C@H]1OC=C[C@@H](O)[C@@H]1OCC1=CC=CC=C1

Specifications

Melting Point 99°C to 103°C
Quantity 1 g
Optical Rotation +10° (c=1 in Chloroform)
Formula Weight 236.26
Percent Purity 97%
Chemical Name or Material 4-O-Benzyl-D-glucal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.