missing translation for 'onlineSavingsMsg'
Learn More

4-Nonylphenol polyethoxylate, MedChemExpress

Product Code. 16492299
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492299 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 16492299 Supplier Medchem Express Supplier No. HYB1027
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

MedChemExpress 4-Nonylphenol polyethoxylate is an organic compound that is a nonionic surfactant; widely used in contraceptives for its spermicidal properties.

4-Nonylphenol polyethoxylate is an organic compound that is a nonionic surfactant

Specifications

Chemical Name or Material 4-Nonylphenol polyethoxylate
CAS 26027-38-3
Color Colorless
Purity Grade Notes Research
Quantity 1 g
Synonym 4-Nonylphenol polyethoxylate Nonoxinol-9 Nonoxynol
Solubility Information Ethanol : 50 mg/mL (Need ultrasonic) ∣DMSO : ≥ 45 mg/mL
SMILES CCCCCCCCCC1=CC=C(OCCO)C=C1.[n]
Percent Purity 98.0%
Grade Research
Physical Form Oil
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.