missing translation for 'onlineSavingsMsg'
Learn More

4-Nonylphenol, 99%, mixture of isomers

Product Code. 10165363 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Glass Bottle
Plastic drum
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10165363 5 kg Plastic drum 5kg
10762022 1 kg Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10165363 Supplier Thermo Scientific Acros Supplier No. 416240050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 84852-15-3 | C15H24O | 220.356 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84852-15-3
MDL Number MFCD00002396
InChI Key SNQQPOLDUKLAAF-UHFFFAOYSA-N
Synonym phenol, 2-nonyl, nonyl phenol, o-nonylphenol, nonyl, phenol,nonyl, 2-n-nonyl-phenol, ortho-n-nonyl-phenol, acmc-209t6o, bidd:er0079, nonylphenol, mixture of isomers
PubChem CID 67296
IUPAC Name 2-nonylphenol
SMILES CCCCCCCCCC1=CC=CC=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 0.9400g/mL
Boiling Point 293.0°C to 297.0°C
Flash Point 141°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic drum
Molecular Formula C15H24O
Refractive Index 1.5105 to 1.5165
Quantity 5 kg
Merck Index 14, 6681
Molecular Weight (g/mol) 220.356
Formula Weight 220.35
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material 4-Nonylphenol, Mixture of isomers
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Nonylphenol, mixture of isomers
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.