missing translation for 'onlineSavingsMsg'
Learn More

4-Nonylphenol (mixture of branched chain isomers) (Technical Grade), TRC

Product Code. 30477478
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477478 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477478 Supplier Toronto Research Chemicals Supplier No. TRCN65043550ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nonylphenol
CAS 84852-15-3
Molecular Formula C15 H24 O
Quantity 50 mL
Synonym Phenol','4-nonyl-','branched, Nonylphenol, Monononylphenol, Nonylphenol (technical), 4-Nonylphenol (mixture of branched chain isomers)
InChI Formula InChI=1S/3C15H24O/c1-5-11(2)12(3)10-13(4)14-6-8-15(16)9-7-14;1-5-11(2)12(3)9-13(4)14-7-6-8-15(16)10-14;1-5-11(2)12(3)10-13(4)14-8-6-7-9-15(14)16/h6-9,11-13,16H,5,10H2,1-4H3;6-8,10-13,16H,5,9H2,1-4H3;6-9,11-13,16H,5,10H2,1-4H3
SMILES CCC(C)C(C)CC(C)c1ccc(O)cc1.CCC(C)C(C)CC(C)c2cccc(O)c2.CCC(C)C(C)CC(C)c3ccccc3O
IUPAC Name 2-(4,5-dimethylheptan-2-yl)phenol;3-(4,5-dimethylheptan-2-yl)phenol;4-(4,5-dimethylheptan-2-yl)phenol
Molecular Weight (g/mol) 204.3
Formula Weight 660.5481
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.