missing translation for 'onlineSavingsMsg'
Learn More

4-Nonyl Phenol, TRC

Product Code. 30481664
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30481664 100 mg
30467254 1 g
30479095 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481664 Supplier Toronto Research Chemicals Supplier No. TRCN650430100MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-40-5
Molecular Formula C15 H24 O
Molecular Weight (g/mol) 220.35
Synonym Phenol, 4-nonyl-, Phenol, p-nonyl- (6CI,8CI), 4-Nonylphenol, 4-n-Nonyl phenol, p-NP, p-Nonylphenol, p-n-Nonylphenol, 4-n-Nonylphenol, 4-Nonylphenol
IUPAC Name 4-nonylphenol
SMILES CCCCCCCCCc1ccc(O)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H24O/c1-2-3-4-5-6-7-8-9-14-10-12-15(16)13-11-14/h10-13,16H,2-9H2,1H3
Formula Weight 220.1827
Percent Purity >95%
Chemical Name or Material 4-Nonylphenol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.