missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrothioanisole, 98%

Product Code. 11489866
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489866 5 g 5g
11499866 25 g 25g
11409876 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489866 Supplier Thermo Scientific Alfa Aesar Supplier No. B20339.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 701-57-5 | C7H7NO2S | 169.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 701-57-5
Molecular Formula C7H7NO2S
Molecular Weight (g/mol) 169.198
MDL Number MFCD00010868
InChI Key NEZGPRYOJVPJKL-UHFFFAOYSA-N
Synonym 4-nitrothioanisole, 1-methylthio-4-nitrobenzene, methyl 4-nitrophenyl sulfide, 1-methylsulfanyl-4-nitrobenzene, benzene, 1-methylthio-4-nitro, methyl 4-nitrophenyl sulphide, 1-methylsulfanyl-4-nitro-benzene, pubchem10631, 4-methylthio nitrobenzene
PubChem CID 96109
IUPAC Name 1-methylsulfanyl-4-nitrobenzene
SMILES CSC1=CC=C(C=C1)[N+](=O)[O-]

Tekniske data

Melting Point 66°C to 72°C
Quantity 5 g
Beilstein 1817928
Formula Weight 169.2
Percent Purity 98%
Chemical Name or Material 4-Nitrothioanisole

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.