missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Nitroquinoline N-oxide, 98+%

Product Code. 11327765
Change view
Click to view available options
Quantity:
250mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327765 250mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11327765 Supplier Alfa Aesar™ Supplier No. L02605.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 56-57-5
Molecular Formula C9H6N2O3
MDL Number MFCD00006738
Quantity 250mg
Beilstein 165756
Synonym 4-nitroquinoline n-oxide, 4-nitroquinoline 1-oxide, 4-nitroquinoline-1-oxide, nitrochin, 4-nitroquinoline-n-oxide, 4-nqo, quinoline, 4-nitro-, 1-oxide, 4-nitroquinoline oxide, 4 nqo, 4-nitroquinolin-1-oxide
InChI Key YHQDZJICGQWFHK-UHFFFAOYSA-N
SMILES [O-][N+](=O)C1=C2C=CC=CC2=[N+]([O-])C=C1
IUPAC Name 4-nitroquinolin-1-ium-1-olate
Molecular Weight (g/mol) 190.16
PubChem CID 5955
ChEBI CHEBI:16907
Formula Weight 190.16
Percent Purity ≥98%
Chemical Name or Material 4-Nitroquinoline N-oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.