missing translation for 'onlineSavingsMsg'
Learn More

4-Nitropyridine N-oxide, 97%

Product Code. 10279603 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10279603 100 g Glass bottle 100g
10012051 5 g Glass bottle 5g
10544151 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10279603 Supplier Thermo Scientific Acros Supplier No. 128921000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1124-33-0 | C5H4N2O3 | 140.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1124-33-0
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.10
MDL Number MFCD00006206
InChI Key RXKNNAKAVAHBNK-UHFFFAOYSA-N
Synonym 4-nitropyridine n-oxide, 4-nitropyridine 1-oxide, 4-nitropyridine-n-oxide, 4-nitropyridine-1-oxide, pyridine, 4-nitro-, 1-oxide, 4-nitropyridin-1-ium-1-olate, p-nitropyridine n-oxide, 4-nitropyridine oxide, 4-nitro-n-oxopyridine, 4-nitro-1-oxido-pyridin-1-ium
PubChem CID 14300
SMILES [O-][N+](=O)C1=CC=[N+]([O-])C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 159°C to 164°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: insoluble
IUPAC Name 4-nitropyridin-1-ium-1-olate
Formula Weight 140.1
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material 4-Nitropyridine N-oxide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Nitropyridine N-oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.