missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophthalic acid, 97%

Product Code. 11314689
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314689 5 g 5g
11324689 25 g 25g
11494173 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11314689 Supplier Thermo Scientific Alfa Aesar Supplier No. A10368.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 610-27-5 | C8H5NO6 | 211.129 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-27-5
Molecular Formula C8H5NO6
Molecular Weight (g/mol) 211.129
MDL Number MFCD00007252
InChI Key SLBQXWXKPNIVSQ-UHFFFAOYSA-N
Synonym 4-nitrophthalic acid, 5-nitrophthalic acid, 4-nitrobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-nitro, phthalic acid, 4-nitro, 4-nitrophthalicacid, p-nitrophthalic acid, 4-nitro-phthalic acid, acmc-209mo8
PubChem CID 69121
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)C(=O)O

Specifications

Melting Point 160°C to 165°C
Odor Odorless
Quantity 5 g
Beilstein 1882262
Formula Weight 211.13
Percent Purity 97%
Chemical Name or Material 4-Nitrophthalic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.