missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenylphosphorylcholine, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
500 mg
1 g
Description
O-(4-Nitrophenylphosphoryl)choline is a useful probe in the assay of Phospholipase C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
O-(4-Nitrophenylphosphoryl)choline is a useful probe in the assay of Phospholipase C.
Solubility
Soluble in water
Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
O-(4-Nitrophenylphosphoryl)choline is a useful probe in the assay of Phospholipase C.
Solubility
Soluble in water
Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Melting Point | 233°C to 235°C |
Color | Yellow |
Quantity | 500 mg |
Beilstein | 1895422 |
Sensitivity | Light and moisture sensitive |
Solubility Information | Soluble in water |
Formula Weight | 304.24 |
Physical Form | Solid |
Chemical Name or Material | 4-Nitrophenylphosphorylcholine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'