missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenylglyoxal hydrate, 97%, Thermo Scientific™

Catalog Number 15437457
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
4974-57-6
C8H5NO4
179.13
MFCD00038682
ILVKBBGIGNSVDO-UHFFFAOYSA-N
4-nitrophenylglyoxal, p-nitrophenylglyoxal, 2-4-nitrophenyl-2-oxoacetaldehyde, para-nitrophenylglyoxal, 4-nitrophenyl glyoxal, 1-4-nitrophenyl glyoxal, 4-nitrophenyl oxo acetaldehyde, 2-4-nitrophenyl-2-oxo-acetaldehyde, benzeneacetaldehyde, 4-nitro-alpha-oxo
151242
2-(4-nitrophenyl)-2-oxoacetaldehyde
[O-][N+](=O)C1=CC=C(C=C1)C(=O)C=O
This item is not returnable. View return policy
4974-57-6
C8H5NO4
179.13
MFCD00038682
ILVKBBGIGNSVDO-UHFFFAOYSA-N
4-nitrophenylglyoxal, p-nitrophenylglyoxal, 2-4-nitrophenyl-2-oxoacetaldehyde, para-nitrophenylglyoxal, 4-nitrophenyl glyoxal, 1-4-nitrophenyl glyoxal, 4-nitrophenyl oxo acetaldehyde, 2-4-nitrophenyl-2-oxo-acetaldehyde, benzeneacetaldehyde, 4-nitro-alpha-oxo
151242
2-(4-nitrophenyl)-2-oxoacetaldehyde
[O-][N+](=O)C1=CC=C(C=C1)C(=O)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
As important industrial compounds and are mainly used as solvents. A new method for arginase assay

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 96°C to 98°C
Color Cream
Quantity 10 g
Formula Weight 179.13 (anhy.)
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenylglyoxal hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'