missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenylacetonitrile, TRC

Product Code. 30473295
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
missing translation for 'unitSize'
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473295 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30473295 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCN50238010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Nitrophenylacetonitrile
CAS 555-21-5
Molecular Formula C8H6N2O2
Quantity 10 g
Synonym (p-Nitrophenyl)-acetonitrile, (p-Nitrophenyl)acetonitrile, 1-(4-Nitrophenyl)acetonitrile, 4-Nitrobenzeneacetonitrile, 4-Nitrobenzyl Cyanide, 4-Nitrobenzyl Nitrile, 4-Nitrophenylacetonitrile, NSC 5396, p-Nitro-alpha-tolunitrile, p-Nitrobenzeneacetonitrile, p-Nitrobenzyl Cyanide
InChI Formula InChI=1S/C8H6N2O2/c9-6-5-7-1-3-8(4-2-7)10(11)12/h1-4H,5H2
SMILES [O-][N+](=O)c1ccc(CC#N)cc1
IUPAC Name 2-(4-nitrophenyl)acetonitrile
Molecular Weight (g/mol) 162.15
Formula Weight 162.0429
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.