missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenylacetonitrile, 98%

Product Code. 11432577
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432577 250 g 250g
11422577 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432577 Supplier Thermo Scientific Alfa Aesar Supplier No. A14821.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 555-21-5 | C8H6N2O2 | 162.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-21-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00007372
InChI Key PXNJGLAVKOXITN-UHFFFAOYSA-N
Synonym 4-nitrophenylacetonitrile, p-nitrophenylacetonitrile, benzeneacetonitrile, 4-nitro, 4-nitrobenzyl cyanide, 2-4-nitrophenyl acetonitrile, p-nitrobenzyl cyanide, p-nitrobenzylcyanide, 4-nitrobenzeneacetonitrile, acetonitrile, p-nitrophenyl, p-nitrophenyl acetonitrile
PubChem CID 68386
IUPAC Name 2-(4-nitrophenyl)acetonitrile
SMILES C1=CC(=CC=C1CC#N)[N+](=O)[O-]

Specifications

Melting Point 114°C to 117°C
Boiling Point 195°C to 197°C (12 mmHg)
Quantity 250 g
UN Number UN3439
Beilstein 388442
Merck Index 14,6590
Formula Weight 162.15
Percent Purity 98%
Chemical Name or Material 4-Nitrophenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.