missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenylacetonitrile, 98%

Product Code. 10056053 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10056053 500 g Glass bottle 500g
10511231 25 g Glass bottle 25g
10488730 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10056053 Supplier Thermo Scientific Acros Supplier No. 166065000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 555-21-5 | C8H6N2O2 | 162.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-21-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00007372
InChI Key PXNJGLAVKOXITN-UHFFFAOYSA-N
Synonym 4-nitrophenylacetonitrile, p-nitrophenylacetonitrile, benzeneacetonitrile, 4-nitro, 4-nitrobenzyl cyanide, 2-4-nitrophenyl acetonitrile, p-nitrobenzyl cyanide, p-nitrobenzylcyanide, 4-nitrobenzeneacetonitrile, acetonitrile, p-nitrophenyl, p-nitrophenyl acetonitrile
PubChem CID 68386
IUPAC Name 2-(4-nitrophenyl)acetonitrile
SMILES C1=CC(=CC=C1CC#N)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 114°C to 118°C
Color Cream to Yellow
Boiling Point 195°C to 197°C (16.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula O2NC6H4CH2CN
Quantity 500 g
Beilstein 09, 456
Merck Index 15, 6678
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alchol,ether and benzene
Formula Weight 162.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenylacetonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 4-Nitrophenylacetonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.