missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenylacetonitrile, 98%, Thermo Scientific Chemicals

Catalog Number 10488730
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
555-21-5
C8H6N2O2
162.15
MFCD00007372
PXNJGLAVKOXITN-UHFFFAOYSA-N
4-nitrophenylacetonitrile, p-nitrophenylacetonitrile, benzeneacetonitrile, 4-nitro, 4-nitrobenzyl cyanide, 2-4-nitrophenyl acetonitrile, p-nitrobenzyl cyanide, p-nitrobenzylcyanide, 4-nitrobenzeneacetonitrile, acetonitrile, p-nitrophenyl, p-nitrophenyl acetonitrile
68386
2-(4-nitrophenyl)acetonitrile
C1=CC(=CC=C1CC#N)[N+](=O)[O-]
This item is not returnable. View return policy
555-21-5
C8H6N2O2
162.15
MFCD00007372
PXNJGLAVKOXITN-UHFFFAOYSA-N
4-nitrophenylacetonitrile, p-nitrophenylacetonitrile, benzeneacetonitrile, 4-nitro, 4-nitrobenzyl cyanide, 2-4-nitrophenyl acetonitrile, p-nitrobenzyl cyanide, p-nitrobenzylcyanide, 4-nitrobenzeneacetonitrile, acetonitrile, p-nitrophenyl, p-nitrophenyl acetonitrile
68386
2-(4-nitrophenyl)acetonitrile
C1=CC(=CC=C1CC#N)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 114°C to 118°C
Color Cream to Yellow
Boiling Point 195°C to 197°C (16.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula O2NC6H4CH2CN
Quantity 100 g
Beilstein 09, 456
Merck Index 15, 6678
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alchol,ether and benzene
Formula Weight 162.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenylacetonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 4-Nitrophenylacetonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'