missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, Thermo Scientific Chemicals

Catalog Number p-4509475
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
333338-18-4
C6H4NNa2O6P
263.05
MFCD00007319
VIYFPAMJCJLZKD-UHFFFAOYSA-L
pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
77949
disodium;(4-nitrophenyl) phosphate
[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1
333338-18-4
C6H4NNa2O6P
263.05
MFCD00007319
VIYFPAMJCJLZKD-UHFFFAOYSA-L
pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
77949
disodium;(4-nitrophenyl) phosphate
[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1

4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, C6H4NNa2O6P.6H2O, CAS Number-333338-18-4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 0.1% max. free 4-nitrophenol (HPLC)
Packaging Glass bottle
Linear Formula O2NC6H4OP(O)(ONa)2·6H2O
Quantity 10 g
Beilstein 06,237
Solubility Information (5% in water) Clear
Formula Weight 371.14
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Nitrophenyl phosphate, disodium salt, hexahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'