missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl isocyanate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4-Nitrophenyl isocyanate is used as pharmaceutical intermediates. 4-Nitrophenyl isocyanate was used in the synthesis of 2-(5-methyl-3,4-diphenyl-1H-pyrrole-2-carbonyl)-N-(4-nitrophenyl)hydrazinecarboxamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrophenyl isocyanate is used as pharmaceutical intermediates. 4-Nitrophenyl isocyanate was used in the synthesis of 2-(5-methyl-3,4-diphenyl-1H-pyrrole-2-carbonyl)-N-(4-nitrophenyl)hydrazinecarboxamide.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Store at 2-8°C. Incompatible with water, heat and oxidizing agents.
4-Nitrophenyl isocyanate is used as pharmaceutical intermediates. 4-Nitrophenyl isocyanate was used in the synthesis of 2-(5-methyl-3,4-diphenyl-1H-pyrrole-2-carbonyl)-N-(4-nitrophenyl)hydrazinecarboxamide.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Store at 2-8°C. Incompatible with water, heat and oxidizing agents.
Specifications
Melting Point | 56°C to 59°C |
Boiling Point | 137°C to 138°C (11 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 1 g |
UN Number | UN3335 |
Beilstein | 389516 |
Formula Weight | 164.12 |
Percent Purity | 97% |
Chemical Name or Material | 4-Nitrophenyl isocyanate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'