missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl isocyanate, 97%

Product Code. 11473494
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473494 5 g 5g
11334689 1 g 1g
11483494 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473494 Supplier Thermo Scientific Alfa Aesar Supplier No. A11194.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-28-7 | C7H4N2O3 | 164.12 g/mol

4-Nitrophenyl isocyanate is used as pharmaceutical intermediates. 4-Nitrophenyl isocyanate was used in the synthesis of 2-(5-methyl-3,4-diphenyl-1H-pyrrole-2-carbonyl)-N-(4-nitrophenyl)hydrazinecarboxamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl isocyanate is used as pharmaceutical intermediates. 4-Nitrophenyl isocyanate was used in the synthesis of 2-(5-methyl-3,4-diphenyl-1H-pyrrole-2-carbonyl)-N-(4-nitrophenyl)hydrazinecarboxamide.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store at 2-8°C. Incompatible with water, heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-28-7
Molecular Formula C7H4N2O3
Molecular Weight (g/mol) 164.12
MDL Number MFCD00007306
InChI Key GFNKTLQTQSALEJ-UHFFFAOYSA-N
Synonym 4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate
PubChem CID 66012
IUPAC Name 1-isocyanato-4-nitrobenzene
SMILES C1=CC(=CC=C1N=C=O)[N+](=O)[O-]

Specifications

Melting Point 56°C to 59°C
Boiling Point 137°C to 138°C (11 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3335
Beilstein 389516
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 164.12
Percent Purity 97%
Chemical Name or Material 4-Nitrophenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.