missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl formate, 97%, Thermo Scientific™

Product Code. p-4258885 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11342640

Brand: Thermo Scientific Acros 438780050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1865-01-6
Molecular Weight (g/mol) 167.12
InChI Key IEXRKQFZXJSHOB-UHFFFAOYSA-N
PubChem CID 74628
IUPAC Name (4-nitrophenyl) formate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 66.0°C to 67.0°C
Color White
Elemental Analysis 48.30 to 52.32% C, 8.05 to 8.72% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H5NO4
Quantity 5 g
Synonym p-nitrophenyl formate, formic acid, 4-nitrophenyl ester, 4-nitrophenyl formate, formic acid, p-nitrophenyl ester, formic acid 4-nitrophenyl ester, acmc-1bty1, formic acid,4-nitrophenyl ester, formic acid 4-nitro-phenyl ester, 4-nitrophenyl formate hplc
Formula Weight 167.12
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Nitrophenyl formate
Show More Show Less
Product Identifier
  • 4-Nitrophenyl formate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.