missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl-beta-D-glucuronide, 99+%

Product Code. 10086393 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10086393 1 g Glass bottle 1g
10623352 50 mg Glass bottle 50mg
10168093 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10086393 Supplier Thermo Scientific Acros Supplier No. 336790010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10344-94-2 | C12H12NO9 | 314.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10344-94-2
Molecular Formula C12H12NO9
Molecular Weight (g/mol) 314.23
MDL Number MFCD00036210
InChI Key QSUILVWOWLUOEU-GOVZDWNOSA-M
Synonym 4-nitrophenyl-beta-d-glucuronide, 4-nitrophenylglucuronide, 4-nitrophenyl beta-d-glucopyranosiduronic acid, 4-nitrophenyl beta-d-glucuronide, p-nitrophenyl beta-d-glucopyranosiduronic acid, 4-nitrophenyl b-d-glucosiduronic acid, p-nitrophenyl-b-d-glucuronide, p-nitrophenyl beta-d-glucuronide, beta-d-glucopyranosiduronic acid, 4-nitrophenyl, labeled with carbon-14, 4-nitrophenyl-beta-d-glucopyranosiduronic acid
PubChem CID 82565
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-nitrophenoxy)oxane-2-carboxylate
SMILES O[C@@H]1[C@@H](O)[C@H](OC2=CC=C(C=C2)[N+]([O-])=O)O[C@@H]([C@H]1O)C([O-])=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 0.05% max. (p-Nitrophenol) (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation −107° to −113° (20°C, 589nm) (c=1, H2O)
Solubility Information (1% in water) Clear colorless to light yellow
Formula Weight 315.24
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 4-Nitrophenyl-β-D-glucuronide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.