missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 4-Nitrophenyl-beta-D-Glucuronide

Product Code. 15825568 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15825568 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15825568 Supplier Thermo Scientific™ Supplier No. J34136.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Nitrophenyl-beta-D-Glucuronide
CAS 10344-94-2
Physical Form Powder
Quantity 1 g
Packaging Plastic Bottle
Synonym 4-nitrophenyl-beta-d-glucuronide
Molecular Weight (g/mol) 315.234
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.