missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl-alpha-L-Fucopyranoside

Product Code. 15895208 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15895208 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15895208 Supplier Thermo Scientific Supplier No. J34131.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10231-84-2285.252 g/mol

 

TRUSTED_SUSTAINABILITY

Specifications

CAS 10231-84-2
Physical Form Powder
Quantity 1 g
Packaging Plastic Bottle
Synonym 4-nitrophenyl a-l-fucopyranoside
Molecular Weight (g/mol) 285.252
Chemical Name or Material 4-Nitrophenyl-alpha-L-Fucopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.