missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl alpha-D-mannopyranoside, 98%

Product Code. 15487219
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15487219 500 mg 500mg
15477219 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487219 Supplier Thermo Scientific Alfa Aesar Supplier No. J66026.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10357-27-4 | C12H15NO8 | 301.251 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10357-27-4
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.251
MDL Number MFCD00066002
InChI Key IFBHRQDFSNCLOZ-GCHJQGSQSA-N
Synonym 4-nitrophenyl a-d-mannopyranoside, 4-nitrophenyl alpha-d-mannopyranoside, 4-nitrophenyl-alpha-d-mannopyranoside, a-d-mannopyranoside, 4-nitrophenyl, p-nitrophenyl-alpha-d-mannopyranoside, 4'-nitrophenyl-alpha-d-mannopyranoside, 2r,3s,4s,5s,6r-2-hydroxymethyl-6-4-nitrophenoxy oxane-3,4,5-triol, pnpalphaman, pnp-alpha-d-man, 4-nitrophenyla-d-mannopyranoside
PubChem CID 449023
IUPAC Name (2R,3S,4S,5S,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 178°C to 183°C
Color White
Quantity 500 mg
Beilstein 92210
Solubility Information Soluble in water at 10mg/ml
Formula Weight 301.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Nitrophenyl alpha-D-mannopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.