missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl-alpha-D-glucopyranoside, 98+%

Product Code. 11302427
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11302427

Brand: Thermo Scientific Alfa Aesar L10070.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3767-28-0 | C12H15NO8 | 301.251 g/mol

4-Nitrophenyl-α-D-glucopyranoside acts as a chromogenic substrate for alfa-D-glucosidase inhibitor. It is also used in the detection of glucansucrases and for yeast alfa-D-glucosidase. Further, it is a substrate for lysosomal alfa-glucosidase and maltase-glucoamylase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl-α-D-glucopyranoside acts as a chromogenic substrate for alfa-D-glucosidase inhibitor. It is also used in the detection of glucansucrases and for yeast alfa-D-glucosidase. Further, it is a substrate for lysosomal alfa-glucosidase and maltase-glucoamylase.

Solubility
Soluble in water, warm ethanol and methanol.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3767-28-0
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.251
MDL Number MFCD00064088
InChI Key IFBHRQDFSNCLOZ-ZIQFBCGOSA-N
Synonym 4-nitrophenyl-alpha-d-glucopyranoside, 4-nitrophenyl alpha-d-glucopyranoside, 4-nitrophenyl a-d-glucopyranoside, 4-nitrophenyl alpha-glucoside, pnpalphaglu, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-4-nitrophenoxy oxane-3,4,5-triol, p-nitrophenyl alpha-d-glucopyranoside, 4'-nitrophenyl-alpha-d-glucopyranoside, png, p-nitrophenyl, a-d-glucopyranoside
PubChem CID 92969
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 206°C to 207°C
Quantity 1 g
Beilstein 92212
Solubility Information Soluble in water,warm ethanol and methanol.
Optical Rotation +218° (c=0.5 in Water)
Formula Weight 301.25
Percent Purity ≥98%
Chemical Name or Material 4-Nitrophenyl-alpha-D-glucopyranoside

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.