missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
4-Nitrophenyl acetate is used with iodoacetic acid for reductive cleavage of methionine-containing peptides. It is also used as a substrate that has been used in assays for esterase and lipase activity. Inorganic complexes have been evaluated for their methanolysis or hydrolysis activity using p-nitrophenyl acetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrophenyl acetate is used with iodoacetic acid for reductive cleavage of methionine-containing peptides. It is also used as a substrate that has been used in assays for esterase and lipase activity. Inorganic complexes have been evaluated for their methanolysis or hydrolysis activity using p-nitrophenyl acetate.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
4-Nitrophenyl acetate is used with iodoacetic acid for reductive cleavage of methionine-containing peptides. It is also used as a substrate that has been used in assays for esterase and lipase activity. Inorganic complexes have been evaluated for their methanolysis or hydrolysis activity using p-nitrophenyl acetate.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 77°C to 79°C |
Quantity | 50 g |
Beilstein | 515874 |
Solubility Information | Insoluble in water. |
Formula Weight | 181.15 |
Percent Purity | ≥98% |
Chemical Name or Material | 4-Nitrophenyl acetate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'