missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford 4-Nitrophenyl-a-L-arabinofuranoside
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 6892-58-6
- Molecular Formula: C11H13NO7
- Molecular Weight: 271.22
Specifications
Specifications
| Quantity | 100 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction