missing translation for 'onlineSavingsMsg'
Learn More

Melford 4-Nitrophenyl-a-L-arabinofuranoside

Product Code. 30018603
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018603 25 mg 25mg
30018605 500 mg 500mg
30018604 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018603 Supplier Melford Supplier No. N660600.025
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • CAS Number: 6892-58-6
  • Molecular Formula: C11H13NO7
  • Molecular Weight: 271.22

Specifications

Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.