missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl 2-(trimethylsilyl)ethyl carbonate, 95%

Product Code. 15445776
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15445776 5 g 5g
15435776 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445776 Supplier Thermo Scientific Alfa Aesar Supplier No. H60466.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80149-80-0 | C12H17NO5Si | 283.36 g/mol

It is employed as a reagent for the introduction of the TEOC-amino protecting group which can be cleaved by F-.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reagent for the introduction of the TEOC-amino protecting group which can be cleaved by F-.

Solubility
Slightly soluble in water.

Notes
Moisture & Light Sensitive. Store at 2-8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80149-80-0
Molecular Formula C12H17NO5Si
Molecular Weight (g/mol) 283.36
MDL Number MFCD00042930
InChI Key ZAQWGGKIMQIVGM-UHFFFAOYSA-N
Synonym 4-nitrophenyl 2-trimethylsilyl ethyl carbonate, teoc-onp, 2-trimethylsilyl ethyl p-nitrophenyl carbonate, teoc-onp;4-nitrophenyl-2-trimethylsilyl ethyl carbonate, 4-nitrophenyl 2-trimethylsilylethyl carbonate, 2-trimethylsilyl ethyl 4-nitrophenyl carbonate, 4-2-trimethylsilyl ethoxycarbonyloxy nitrobenzene, carbonic acid, 4-nitrophenyl 2-trimethylsilyl ethyl ester, ambotzrl-8204
PubChem CID 3086116
IUPAC Name (4-nitrophenyl) 2-trimethylsilylethyl carbonate
SMILES C[Si](C)(C)CCOC(=O)OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 35°C to 40°C
Linear Formula C12H17NO5Si
Quantity 5 g
Beilstein 4320585
Sensitivity Light and moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 283.35
Percent Purity 95%
Chemical Name or Material 4-Nitrophenyl 2-(trimethylsilyl)ethyl carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.