Learn More
Melford 4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside
Product Code. 30018593
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 30018593
Brand: Melford N516000.25
Chromogenic substrate for detecting N-Acetyl-β-D-glucosaminidase.
Chromogenic substrate for detecting N-Acetyl-β-D-glucosaminidase.
Specifications
Quantity 250 mg