missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl 2-(Acetamido)-2-deoxy-3-O-Beta-D-galactopyranosyl-Alpha-D-galactopyranoside, TRC

Product Code. 30464671
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30464671 1 mg
30483093 10 mg
30467257 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30464671 Supplier Toronto Research Chemicals Supplier No. TRCN5010001MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59837-14-8
Molecular Formula C20 H28 N2 O13
Molecular Weight (g/mol) 504.44
IUPAC Name N-[(2R,3R,4R,5R,6R)-5-hydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)-4-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-3-yl]acetamide
SMILES CC(=O)N[C@H]1[C@@H](Oc2ccc(cc2)[N+](=O)[O-])O[C@H](CO)[C@H](O)[C@@H]1O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H28N2O13/c1-8(25)21-13-18(35-20-17(29)16(28)14(26)11(6-23)34-20)15(27)12(7-24)33-19(13)32-10-4-2-9(3-5-10)22(30)31/h2-5,11-20,23-24,26-29H,6-7H2,1H3,(H,21,25)/t11-,12-,13-,14+,15+,16+,17-,18-,19+,20+/m1/s1
Formula Weight 504.1591
Percent Purity >95%
Chemical Name or Material 4-Nitrophenyl 2-(Acetamido)-2-deoxy-3-O-beta-D-galactopyranosyl-alpha-D-galactopyranoside
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.