missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenol, TRC

Product Code. 30135387
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135387 100 g 100g
18001920 10 g 10g
18013020 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135387 Supplier Toronto Research Chemicals Supplier No. TRCN496945100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-02-7
Formula Weight 139.0269
InChI Formula InChI=1S/C6H5NO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H
IUPAC Name 4-nitrophenol
Molecular Formula C6 H5 N O3
Molecular Weight (g/mol) 139.11
Percent Purity >95%
SMILES Oc1ccc(cc1)[N+](=O)[O-]
Synonym 4-Nitrophenol, Paracetamol Imp. F (EP), Phenol, 4-nitro- (9CI, ACI), 4-Nitrophenol (ACI), Phenol, p-nitro- (8CI), 1-Hydroxy-4-nitrobenzene, 4-Hydroxy-1-nitrobenzene, 4-Hydroxynitrobenzene, Niphen, NSC 1317, p-Hydroxynitrobenzene, p-Nitrophenol
Chemical Name or Material 4-Nitrophenol
Grade HPLC
Show More Show Less

Specifications

Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.